Product Details

SNP ID
rs146332668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132843016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCCCCAGCTGCACTCACATGGC[A/G]TGGTGAGCTTTCACCTGAGTGCGGC
Phenotype
MIM: 605209
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHFR PubMed Links

Gene Details

Gene
CHFR
Gene Name
checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161344.1 1908 Silent Mutation CAC,CAT H649H NP_001154816.1
NM_001161345.1 1908 Silent Mutation CAC,CAT H648H NP_001154817.1
NM_001161346.1 1908 Silent Mutation CAC,CAT H637H NP_001154818.1
NM_001161347.1 1908 Silent Mutation CAC,CAT H557H NP_001154819.1
NM_018223.2 1908 Silent Mutation CAC,CAT H608H NP_060693.2

View Full Product Details