Product Details

SNP ID
rs146486585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6200544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGTGCATCAAATACCTGCTGTT[A/C]GGATTTAACTTCATCTTCTGGGTGA
Phenotype
MIM: 143030
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD9 PubMed Links

Gene Details

Gene
CD9
Gene Name
CD9 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001769.3 229 Missense Mutation TTA,TTC L15F NP_001760.1
XM_005253814.3 229 Missense Mutation TTA,TTC L15F XP_005253871.1
XM_011521040.1 229 Intron XP_011519342.1

View Full Product Details