Product Details

SNP ID
rs146722647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48184450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGGCGTGGTCGGCGGCAAGGGG[A/G]GCTTCCAGAGCGACCCCCAAGAGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC184 PubMed Links

Gene Details

Gene
CCDC184
Gene Name
coiled-coil domain containing 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013635.3 868 Missense Mutation AGC,GGC S110G NP_001013657.3

View Full Product Details