Product Details

SNP ID
rs146955896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108909173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTTCTTGATAACATTGTTGAGC[A/G]TTGAACCTTCTCTGATCTCTGCCTG
Phenotype
MIM: 124050 MIM: 611699
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAO PubMed Links

Gene Details

Gene
DAO
Gene Name
D-amino acid oxidase
There are no transcripts associated with this gene.

Gene
SVOP
Gene Name
SV2 related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018711.4 5381 UTR 3 NP_061181.1

View Full Product Details