Product Details

SNP ID
rs147261688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49089667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTAAGGAGTGCGCACCATCAGC[C/G]CTACCTACCTAGGACCCGGTATCAC
Phenotype
MIM: 605423 MIM: 610007
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DHH PubMed Links

Gene Details

Gene
DHH
Gene Name
desert hedgehog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021044.3 1690 UTR 3 NP_066382.1
XM_017019380.1 1690 UTR 3 XP_016874869.1
XM_017019381.1 1690 UTR 3 XP_016874870.1
Gene
LMBR1L
Gene Name
limb development membrane protein 1 like
There are no transcripts associated with this gene.

View Full Product Details