Product Details

SNP ID
rs147339942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:114355566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGCTCCACTCTGGCACCATGCCA[A/G]CTCCGTGCACAGAGTGGTACTGATG
Phenotype
MIM: 601620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBX5 PubMed Links

Gene Details

Gene
TBX5
Gene Name
T-box 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000192.3 3941 Missense Mutation GCT,GTT A508V NP_000183.2
NM_080717.2 3941 Missense Mutation GCT,GTT A458V NP_542448.1
NM_181486.2 3941 Missense Mutation GCT,GTT A508V NP_852259.1
XM_017019912.1 3941 Missense Mutation GCT,GTT A524V XP_016875401.1

View Full Product Details