Product Details

SNP ID
rs147938337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15109182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGGTGGAGCTGCGTCGCCTCGT[C/T]TTGCCCTGCACCATCCTCCGGCGAC
Phenotype
MIM: 612664
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RERG PubMed Links

Gene Details

Gene
RERG
Gene Name
RAS like estrogen regulated growth inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190726.1 868 Silent Mutation AAA,AAG K157K NP_001177655.1
NM_032918.2 868 Silent Mutation AAA,AAG K176K NP_116307.1
XM_017020121.1 868 Silent Mutation AAA,AAG K173K XP_016875610.1

View Full Product Details