Product Details

SNP ID
rs148012469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:280371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCAACACAATTTTTTTTTTTAA[C/T]GGACTTGCCACCTTAAGAAACTTCA
Phenotype
MIM: 180202
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM5A PubMed Links

Gene Details

Gene
KDM5A
Gene Name
lysine demethylase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042603.2 10522 UTR 3 NP_001036068.1

View Full Product Details