Product Details

SNP ID
rs148140770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108524469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATCTTCATCACAGCCTGCGAC[A/G]CCTGGGATTCATTTTCATACTCCAC
Phenotype
MIM: 611684
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FICD PubMed Links

Gene Details

Gene
FICD
Gene Name
FIC domain containing
There are no transcripts associated with this gene.

Gene
SART3
Gene Name
squamous cell carcinoma antigen recognized by T-cells 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014706.3 2331 Missense Mutation GCG,GTG A854V NP_055521.1
XM_005269241.4 2331 Missense Mutation GCG,GTG A872V XP_005269298.1
XM_011539026.2 2331 Missense Mutation GCG,GTG A566V XP_011537328.1

View Full Product Details