Product Details

SNP ID
rs148575498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49089894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTCCGCTAAGCGGTAGAGGA[A/G]CCGAGAGTACCAATGCATGCCAGTC
Phenotype
MIM: 605423 MIM: 610007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHH PubMed Links

Gene Details

Gene
DHH
Gene Name
desert hedgehog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021044.3 1463 Missense Mutation CTC,TTC L386F NP_066382.1
XM_017019380.1 1463 Missense Mutation CTC,TTC L339F XP_016874869.1
XM_017019381.1 1463 Missense Mutation CTC,TTC L272F XP_016874870.1
Gene
LMBR1L
Gene Name
limb development membrane protein 1 like
There are no transcripts associated with this gene.

View Full Product Details