Product Details

SNP ID
rs148698440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:40393462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCACAACCATGAAGTTGATCTT[A/G]TGGTACTTGGTTGTGGCACTTTGGT
Phenotype
MIM: 612170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUC19 PubMed Links

Gene Details

Gene
MUC19
Gene Name
mucin 19, oligomeric
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173600.2 Intron NP_775871.2

View Full Product Details