Product Details

SNP ID
rs149150271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108523596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCACTTGGGCGCTGCAGGGCA[C/T]GAGGCAGTAGAGACAGCTGCGTCCT
Phenotype
MIM: 611684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FICD PubMed Links

Gene Details

Gene
FICD
Gene Name
FIC domain containing
There are no transcripts associated with this gene.

Gene
SART3
Gene Name
squamous cell carcinoma antigen recognized by T-cells 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014706.3 2523 Missense Mutation CAT,CGT H918R NP_055521.1
XM_005269241.4 2523 Missense Mutation CAT,CGT H936R XP_005269298.1
XM_011539026.2 2523 Missense Mutation CAT,CGT H630R XP_011537328.1

View Full Product Details