Product Details

SNP ID
rs149958490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49973362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTACAGATTGCCATCACCTTCAA[C/T]CTGGTCACCGCCATGGCTGTGCAGG
Phenotype
MIM: 600442 MIM: 601383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQP5 PubMed Links

Gene Details

Gene
AQP5
Gene Name
aquaporin 5
There are no transcripts associated with this gene.

Gene
AQP6
Gene Name
aquaporin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001652.3 526 Silent Mutation AAC,AAT N63N NP_001643.2

View Full Product Details