Product Details

SNP ID
rs150183934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15109371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCAGCTTTGTTTCCAACCAAGAT[A/G]AGAGTCACATTCTTGGGCTTTTTGA
Phenotype
MIM: 612664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RERG PubMed Links

Gene Details

Gene
RERG
Gene Name
RAS like estrogen regulated growth inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190726.1 679 Silent Mutation NP_001177655.1
NM_032918.2 679 Silent Mutation NP_116307.1
XM_017020121.1 679 Silent Mutation XP_016875610.1

View Full Product Details