Product Details

SNP ID
rs150351122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21472781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTAAAAGAATTTTAAGTAAAT[A/C]TCCGATGACTAGCCATTAACTGGAT
Phenotype
MIM: 600537
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PYROXD1 PubMed Links

Gene Details

Gene
PYROXD1
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
There are no transcripts associated with this gene.

Gene
RECQL
Gene Name
RecQ like helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002907.3 Intron NP_002898.2
NM_032941.2 Intron NP_116559.1
XM_005253461.3 Intron XP_005253518.1
XM_005253462.4 Intron XP_005253519.1
XM_005253463.3 Intron XP_005253520.1
XM_005253464.3 Intron XP_005253521.1

View Full Product Details