Product Details

SNP ID
rs150469463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64459542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGCCTTTACTCCTCTAGAGAGG[G/T]TCTGTGAAGGAACAGCCTGCCCTGG
Phenotype
MIM: 604834 MIM: 603180
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TBK1 PubMed Links

Gene Details

Gene
TBK1
Gene Name
TANK binding kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013254.3 Intron NP_037386.1
XM_005268809.1 Intron XP_005268866.1
XM_005268810.1 Intron XP_005268867.1
Gene
XPOT
Gene Name
exportin for tRNA
There are no transcripts associated with this gene.

View Full Product Details