Product Details

SNP ID
rs150477287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14914746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCAAGGAGAAGTCAGAGTTCCA[A/C]CTTTGGAGTCTTTCCTTCTGATTCA
Phenotype
MIM: 610642
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ERP27 PubMed Links

Gene Details

Gene
ERP27
Gene Name
endoplasmic reticulum protein 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300784.1 852 Missense Mutation GTG,TTG V170L NP_001287713.1
NM_152321.3 852 Missense Mutation GTG,TTG V271L NP_689534.1

View Full Product Details