Product Details

SNP ID
rs150800253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95518258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTAGAATGTCCATTCTCAGTAA[A/C]TCGTTGGGTATAAAACAAGATATAA
Phenotype
MIM: 601870 MIM: 610993
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
METAP2 PubMed Links

Gene Details

Gene
METAP2
Gene Name
methionyl aminopeptidase 2
There are no transcripts associated with this gene.

Gene
USP44
Gene Name
ubiquitin specific peptidase 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042403.2 2313 Missense Mutation NP_001035862.1
NM_001278393.1 2313 Missense Mutation NP_001265322.1
NM_032147.4 2313 Missense Mutation NP_115523.2
XM_005269172.1 2313 Missense Mutation XP_005269229.1
XM_005269173.1 2313 Missense Mutation XP_005269230.1
XM_005269174.1 2313 Missense Mutation XP_005269231.1
XM_011538800.2 2313 Missense Mutation XP_011537102.1
XM_011538801.1 2313 Missense Mutation XP_011537103.1
XM_011538802.1 2313 Missense Mutation XP_011537104.1
XM_011538803.1 2313 Missense Mutation XP_011537105.1
XM_011538804.2 2313 Missense Mutation XP_011537106.1
XM_011538805.1 2313 Missense Mutation XP_011537107.1
XM_011538806.2 2313 Intron XP_011537108.1
XM_017020009.1 2313 Missense Mutation XP_016875498.1
XM_017020010.1 2313 Missense Mutation XP_016875499.1
XM_017020011.1 2313 Missense Mutation XP_016875500.1
XM_017020012.1 2313 Intron XP_016875501.1

View Full Product Details