Product Details

SNP ID
rs151100410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:18081261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCATTGATTTAGATCCACTGGGA[C/T]GTCTCTTTTCTTTGAGTTTGAAGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RERGL PubMed Links

Gene Details

Gene
RERGL
Gene Name
RERG like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286201.1 755 Missense Mutation CAT,CGT H182R NP_001273130.1
NM_024730.3 755 Missense Mutation CAT,CGT H183R NP_079006.1

View Full Product Details