Product Details

SNP ID
rs151118817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56229110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCCTCCCACCCACCCAGCACCC[A/G]AATCACTCGAAGCCAGCCCAACCAC
Phenotype
MIM: 612104 MIM: 608730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD52 PubMed Links

Gene Details

Gene
ANKRD52
Gene Name
ankyrin repeat domain 52
There are no transcripts associated with this gene.

Gene
NABP2
Gene Name
nucleic acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024068.3 677 Missense Mutation CAA,CGA Q178R NP_076973.1
XM_005269146.2 677 Missense Mutation CAA,CGA Q221R XP_005269203.1
XM_005269147.2 677 Missense Mutation CAA,CGA Q194R XP_005269204.1
XM_005269148.4 677 Missense Mutation CAA,CGA Q178R XP_005269205.1
XM_005269149.4 677 Missense Mutation CAA,CGA Q178R XP_005269206.1
Gene
RNF41
Gene Name
ring finger protein 41
There are no transcripts associated with this gene.

Gene
SLC39A5
Gene Name
solute carrier family 39 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135195.1 677 Intron NP_001128667.1
NM_173596.2 677 Intron NP_775867.2
XM_005268803.1 677 Intron XP_005268860.1
XM_011538198.1 677 Intron XP_011536500.1
XM_011538199.1 677 Intron XP_011536501.1
XM_011538200.1 677 Intron XP_011536502.1
XM_011538201.1 677 Intron XP_011536503.1
XM_017019185.1 677 Intron XP_016874674.1
XM_017019186.1 677 Intron XP_016874675.1
XM_017019187.1 677 Intron XP_016874676.1

View Full Product Details