Product Details

SNP ID
rs151309160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56995927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCTCATCACAGTCTTCAATGTG[C/T]TGACAGCCTGCCGGCTGCGGCAGCC
Phenotype
MIM: 605307
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR182 PubMed Links

Gene Details

Gene
GPR182
Gene Name
G protein-coupled receptor 182
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007264.3 1488 Silent Mutation CTG,TTG L240L NP_009195.1
XM_011537824.2 1488 Missense Mutation CTG,TTG L240L XP_011536126.1
XM_011537825.2 1488 Missense Mutation CTG,TTG L240L XP_011536127.1
XM_011537827.2 1488 Missense Mutation CTG,TTG L240L XP_011536129.1
Gene
HBCBP
Gene Name
HBcAg-binding protein
There are no transcripts associated with this gene.

Gene
ZBTB39
Gene Name
zinc finger and BTB domain containing 39
There are no transcripts associated with this gene.

View Full Product Details