Product Details

SNP ID
rs137963049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99965146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGCAATGTACAAGTCCAGCTT[C/G]GTTGTTTTTTGTTTGTTTTTTAAGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927437 PubMed Links

Gene Details

Gene
LOC101927437
Gene Name
uncharacterized LOC101927437
There are no transcripts associated with this gene.

Gene
ZIC5
Gene Name
Zic family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033132.3 2472 UTR 3 NP_149123.2

View Full Product Details