Product Details
- SNP ID
-
rs138243017
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:70107457 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAGGAAGAGGAGGAAGAGGACGG[A/G]GAGGACGATGAAGAGGAAGAGGAGG
- Phenotype
-
MIM: 603680
MIM: 605332
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN8OS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3751427] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATXN8OS
- Gene Name
- ATXN8 opposite strand (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- KLHL1
- Gene Name
- kelch like family member 1
View Full Product Details