Product Details

SNP ID
rs142944522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49532998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAATAGGATCTTTTCAGGAACA[C/T]CATAATTTTGGTGCTTTTTCAAGGT
Phenotype
MIM: 607796 MIM: 607867
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHF11 PubMed Links

Gene Details

Gene
PHF11
Gene Name
PHD finger protein 11
There are no transcripts associated with this gene.

Gene
RCBTB1
Gene Name
RCC1 and BTB domain containing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018191.3 2907 UTR 3 NP_060661.3
XM_005266441.2 2907 UTR 3 XP_005266498.1
XM_011535133.1 2907 UTR 3 XP_011533435.1
XM_011535134.1 2907 UTR 3 XP_011533436.1
XM_011535135.2 2907 UTR 3 XP_011533437.1
XM_011535136.2 2907 Intron XP_011533438.1
Gene
SETDB2-PHF11
Gene Name
SETDB2-PHF11 readthrough
There are no transcripts associated with this gene.

View Full Product Details