Product Details

SNP ID
rs143109873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102769000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCATATTTATACATGCATAGTA[C/T]GGTAGATGAAATATTCTAGAATGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
There are no transcripts associated with this gene.

Gene
TEX30
Gene Name
testis expressed 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286775.1 206 Intron NP_001273704.1
NM_001286776.1 206 Intron NP_001273705.1
NM_138779.4 206 Intron NP_620134.3
XM_005254097.2 206 Intron XP_005254154.1
XM_011521135.2 206 Intron XP_011519437.1
XM_011521136.2 206 Missense Mutation CAT,CGT H35R XP_011519438.1
XM_017020852.1 206 Intron XP_016876341.1

View Full Product Details