Product Details

SNP ID
rs143268770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:38966986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGAGAGCTATGGGAGACTCAG[C/G]CAGCACCATGGAGGCTGACTTCAGG
Phenotype
MIM: 608327
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STOML3 PubMed Links

Gene Details

Gene
STOML3
Gene Name
stomatin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144033.1 937 Missense Mutation CCT,GCT P230A NP_001137505.1
NM_145286.2 937 Missense Mutation CCT,GCT P239A NP_660329.1
XM_017020400.1 937 Missense Mutation CCT,GCT P168A XP_016875889.1

View Full Product Details