Product Details
- SNP ID
-
rs144343396
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:51865526 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGCACAAGCTGCAGGAGGAAAAG[C/G]CTTTTCGCGAAGAGATGAAAATTTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC70
PubMed Links
Gene Details
- Gene
- CCDC70
- Gene Name
- coiled-coil domain containing 70
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031290.2 |
444 |
Missense Mutation |
CCT,GCT |
P50A |
NP_112580.2 |
View Full Product Details