Product Details

SNP ID
rs147042049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:112068448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACAGCCCCATCTCCAACTCGCAG[A/G]GCTACATGAGCGCGTCGCCCTCGGG
Phenotype
MIM: 602148
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00403 PubMed Links

Gene Details

Gene
LINC00403
Gene Name
long intergenic non-protein coding RNA 403
There are no transcripts associated with this gene.

Gene
SOX1
Gene Name
SRY-box 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005986.2 850 Missense Mutation AGC,GGC S264G NP_005977.2

View Full Product Details