Product Details

SNP ID
rs149886487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:38966893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAATGCCCTCTAGTATATTCA[C/T]GGGCAGAGGAAACACAATCGTAGAA
Phenotype
MIM: 608327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STOML3 PubMed Links

Gene Details

Gene
STOML3
Gene Name
stomatin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144033.1 1030 Missense Mutation ATG,GTG M261V NP_001137505.1
NM_145286.2 1030 Missense Mutation ATG,GTG M270V NP_660329.1
XM_017020400.1 1030 Missense Mutation ATG,GTG M199V XP_016875889.1

View Full Product Details