Product Details

SNP ID
rs150440372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52377634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCTCTGGTAGTTATCTTTGGGG[G/T]ATATGGGAGAAGACTGCTTCCTTGA
Phenotype
MIM: 616821
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
THSD1 PubMed Links

Gene Details

Gene
THSD1
Gene Name
thrombospondin type 1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018676.3 2881 Missense Mutation NP_061146.1
NM_199263.2 2881 Missense Mutation NP_954872.1

View Full Product Details