Product Details

SNP ID
rs150476227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99965740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGGGCTTGTCACTGGTGTGGAC[A/G]TGGGAATGTTTCTTCCGATCACTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927437 PubMed Links

Gene Details

Gene
LOC101927437
Gene Name
uncharacterized LOC101927437
There are no transcripts associated with this gene.

Gene
ZIC5
Gene Name
Zic family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033132.3 1878 Silent Mutation CAC,CAT H543H NP_149123.2

View Full Product Details