Product Details

SNP ID
rs139147634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:61709632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGATAATAATGATCTAAACCAA[G/T]AAATTTAGTTGCTTTCAAAAATAAA
Phenotype
MIM: 603348
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HIF1A PubMed Links

Gene Details

Gene
HIF1A
Gene Name
hypoxia inducible factor 1 alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243084.1 Intron NP_001230013.1
NM_001530.3 Intron NP_001521.1
NM_181054.2 Intron NP_851397.1

View Full Product Details