Product Details

SNP ID
rs139506803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94377035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCAGTGAGTAAGCTTAAGAGAA[C/T]AGGAGACTTGTGTGGGAAACAGTCG
Phenotype
MIM: 107400
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINA1 PubMed Links

Gene Details

Gene
SERPINA1
Gene Name
serpin family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000295.4 3204 UTR 3 NP_000286.3
NM_001002235.2 3204 UTR 3 NP_001002235.1
NM_001002236.2 3204 UTR 3 NP_001002236.1
NM_001127700.1 3204 UTR 3 NP_001121172.1
NM_001127701.1 3204 UTR 3 NP_001121173.1
NM_001127702.1 3204 UTR 3 NP_001121174.1
NM_001127703.1 3204 UTR 3 NP_001121175.1
NM_001127704.1 3204 UTR 3 NP_001121176.1
NM_001127705.1 3204 UTR 3 NP_001121177.1
NM_001127706.1 3204 UTR 3 NP_001121178.1
NM_001127707.1 3204 UTR 3 NP_001121179.1
XM_017021370.1 3204 Intron XP_016876859.1

View Full Product Details