Product Details

SNP ID
rs140038394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94304490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGTCGAAGATCATGATGATGAA[C/G]GGCTGGTTGAAACGCAAGATGATAG
Phenotype
MIM: 122500
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SERPINA6 PubMed Links

Gene Details

Gene
SERPINA6
Gene Name
serpin family A member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001756.3 1250 Silent Mutation CCC,CCG P382P NP_001747.2

View Full Product Details