Product Details

SNP ID
rs141526392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22565222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCATCTAAATTCTAACCCACA[A/T]GACAGTTATGGGGTTCCCACACTCA
Phenotype
MIM: 600243
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DAD1 PubMed Links

Gene Details

Gene
DAD1
Gene Name
defender against cell death 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001344.3 Intron NP_001335.1
Gene
LOC105370399
Gene Name
uncharacterized LOC105370399
There are no transcripts associated with this gene.

View Full Product Details