Product Details

SNP ID
rs141768302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58433647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACCACCTTTTTGTATCAAATTA[C/T]AGCATATGGAAATTTGAGGACAAAG
Phenotype
MIM: 610178 MIM: 607384
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0586 PubMed Links

Gene Details

Gene
KIAA0586
Gene Name
KIAA0586
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244189.1 Intron NP_001231118.1
NM_001244190.1 Intron NP_001231119.1
NM_001244191.1 Intron NP_001231120.1
NM_001244192.1 Intron NP_001231121.1
NM_001244193.1 Intron NP_001231122.1
Gene
TIMM9
Gene Name
translocase of inner mitochondrial membrane 9
There are no transcripts associated with this gene.

View Full Product Details