Product Details

SNP ID
rs141912564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70522755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCACTATCAGCACTACCTCCA[C/T]AGCCTTTGTCCTTGCAGTATGGGGG
Phenotype
MIM: 603712
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM20 PubMed Links

Gene Details

Gene
ADAM20
Gene Name
ADAM metallopeptidase domain 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003814.4 11294 Missense Mutation TAT,TGT Y718C NP_003805.3
XM_005268151.3 11294 Missense Mutation TAT,TGT Y718C XP_005268208.1

View Full Product Details