Product Details

SNP ID
rs143514872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77377616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCGAGGAAGAGGACTTCAAAGA[C/G]GGGGAGCCAGACAGTGCTAAGAACG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SAMD15 PubMed Links

Gene Details

Gene
SAMD15
Gene Name
sterile alpha motif domain containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010860.1 198 Silent Mutation GAC,GAG D66E NP_001010860.1
Gene
TMED8
Gene Name
transmembrane p24 trafficking protein family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213601.1 198 Intron NP_998766.1
XM_017021223.1 198 Intron XP_016876712.1
XM_017021224.1 198 Intron XP_016876713.1
XM_017021225.1 198 Intron XP_016876714.1

View Full Product Details