Product Details

SNP ID
rs143842518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77394757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTACTTGAGAGATGCTGGCTAAA[C/T]GGAGTTTCCTTGAGTTGCACAGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOXRED1 PubMed Links

Gene Details

Gene
NOXRED1
Gene Name
NADP dependent oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113475.2 1034 Silent Mutation CCA,CCG P318P NP_001106946.1
XM_005267330.3 1034 Silent Mutation CCA,CCG P318P XP_005267387.1
XM_011536427.2 1034 Silent Mutation CCA,CCG P318P XP_011534729.1
XM_011536428.2 1034 Silent Mutation CCA,CCG P318P XP_011534730.1
XM_011536429.2 1034 Silent Mutation CCA,CCG P228P XP_011534731.1
XM_017020969.1 1034 Silent Mutation CCA,CCG P193P XP_016876458.1
XM_017020970.1 1034 Intron XP_016876459.1
Gene
SAMD15
Gene Name
sterile alpha motif domain containing 15
There are no transcripts associated with this gene.

View Full Product Details