Product Details

SNP ID
rs143980244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20458637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCCCAAACACAGCACAGCCAA[C/G]AGGATGACAAATGCCCAGGCTGCAT
Phenotype
MIM: 107748 MIM: 610107 MIM: 609865
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APEX1 PubMed Links

Gene Details

Gene
APEX1
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244249.1 1064 Intron NP_001231178.1
NM_001641.3 1064 Intron NP_001632.2
NM_080648.2 1064 Intron NP_542379.1
NM_080649.2 1064 Intron NP_542380.1
Gene
OSGEP
Gene Name
O-sialoglycoprotein endopeptidase
There are no transcripts associated with this gene.

Gene
TMEM55B
Gene Name
transmembrane protein 55B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100814.2 1064 Silent Mutation CTC,CTG L259L NP_001094284.1
NM_144568.3 1064 Silent Mutation CTC,CTG L252L NP_653169.2

View Full Product Details