Product Details

SNP ID
rs144904034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:51495503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGCTCTTCTGGCAAATTGCCAGA[C/T]TGATTTCCTCAGAAACCAGCAACAA
Phenotype
MIM: 614555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRMD6 PubMed Links

Gene Details

Gene
FRMD6
Gene Name
FERM domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042481.2 Intron NP_001035946.1
NM_001267046.1 Intron NP_001253975.1
NM_001267047.1 Intron NP_001253976.1
NM_152330.3 Intron NP_689543.1
XM_006720029.1 Intron XP_006720092.1
XM_006720030.1 Intron XP_006720093.1
XM_011536423.1 Intron XP_011534725.1
XM_011536424.1 Intron XP_011534726.1
XM_017020967.1 Intron XP_016876456.1
XM_017020968.1 Intron XP_016876457.1
Gene
FRMD6-AS2
Gene Name
FRMD6 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details