Product Details

SNP ID
rs145938782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34712925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTCCCAATTTCTCTCCAAGTGT[C/T]GAACGGTCCTTAATATCATCCAAGC
Phenotype
MIM: 601443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFL2 PubMed Links

Gene Details

Gene
CFL2
Gene Name
cofilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243645.1 520 Silent Mutation NP_001230574.1
NM_021914.7 520 Silent Mutation NP_068733.1
NM_138638.4 520 Silent Mutation NP_619579.1
XM_011536363.2 520 Silent Mutation XP_011534665.1

View Full Product Details