Product Details

SNP ID
rs146867283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104586477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGCCCTGACTCCCGGCCAGAGA[C/T]ACGACGTCAGACCAGAAAGAATGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf180 PubMed Links

Gene Details

Gene
C14orf180
Gene Name
chromosome 14 open reading frame 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008404.2 383 Missense Mutation ACA,ATA T16I NP_001008404.1
NM_001286399.1 383 Missense Mutation ACA,ATA T16I NP_001273328.1
NM_001286400.1 383 Missense Mutation ACA,ATA T16I NP_001273329.1
XM_005267638.3 383 Missense Mutation ACA,ATA T16I XP_005267695.1
XM_011536764.2 383 Missense Mutation ACA,ATA T16I XP_011535066.1
Gene
TMEM179
Gene Name
transmembrane protein 179
There are no transcripts associated with this gene.

View Full Product Details