Product Details

SNP ID
rs147453957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23357508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCTTGCTGTGGGGCACGAAAAG[G/T]CGCGGGGGCTGATTAGCCTGGGTAC
Phenotype
MIM: 609906 MIM: 611461
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFS PubMed Links

Gene Details

Gene
EFS
Gene Name
embryonal Fyn-associated substrate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277174.1 1505 Silent Mutation CGA,CGC R299R NP_001264103.1
NM_005864.3 1505 Silent Mutation CGA,CGC R468R NP_005855.1
NM_032459.2 1505 Silent Mutation CGA,CGC R375R NP_115835.1
XM_005267256.1 1505 Silent Mutation CGA,CGC R438R XP_005267313.1
Gene
SLC22A17
Gene Name
solute carrier family 22 member 17
There are no transcripts associated with this gene.

View Full Product Details