Product Details

SNP ID
rs148308026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91114479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAGCTGTTCCTCTCCTTGAAA[A/T]GTCAAGGCTTGGTTCAAGCCAGATA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C14orf159 PubMed Links

Gene Details

Gene
C14orf159
Gene Name
chromosome 14 open reading frame 159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102366.2 467 UTR 5 NP_001095836.1
NM_001102367.2 467 Intron NP_001095837.1
NM_001102368.2 467 Intron NP_001095838.1
NM_001102369.2 467 Intron NP_001095839.1
NM_001286470.1 467 UTR 5 NP_001273399.1
NM_001286471.1 467 Intron NP_001273400.1
NM_001286472.1 467 Intron NP_001273401.1
NM_001286473.1 467 Intron NP_001273402.1
NM_024952.7 467 UTR 5 NP_079228.4

View Full Product Details