Product Details

SNP ID
rs148447482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:61766974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCATACACCTTCCAGATCAGAG[C/T]TGGTGCTTTGTATCTGCTATATGGA
Phenotype
MIM: 600591
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNAPC1 PubMed Links

Gene Details

Gene
SNAPC1
Gene Name
small nuclear RNA activating complex polypeptide 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003082.3 331 Missense Mutation GCT,GTT A76V NP_003073.1

View Full Product Details