Product Details

SNP ID
rs148898725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22875527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCGTCCCCTCCCTGCCTTGCAA[C/T]GTCACCTTGGAGGACTTCTATGGGG
Phenotype
MIM: 609921 MIM: 616955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP10 PubMed Links

Gene Details

Gene
LRP10
Gene Name
LDL receptor related protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005267510.1 1294 Silent Mutation AAC,AAT N201N XP_005267567.1
Gene
REM2
Gene Name
RRAD and GEM like GTPase 2
There are no transcripts associated with this gene.

View Full Product Details