Product Details

SNP ID
rs149752323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23118789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCTGTAGATGCCAGGCCCCAG[C/T]GCCTTCCTGTCTGGGCCGAAGGTAT
Phenotype
MIM: 600749 MIM: 604235
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEBPE PubMed Links

Gene Details

Gene
CEBPE
Gene Name
CCAAT/enhancer binding protein epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001805.3 823 Silent Mutation GCA,GCG A101A NP_001796.2
Gene
SLC7A8
Gene Name
solute carrier family 7 member 8
There are no transcripts associated with this gene.

View Full Product Details