Product Details

SNP ID
rs150810487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:31483598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGTGAGAAGGTAACGATCAAGA[C/T]CGATGGCCGAAAGGAAGAAAACAGA
Phenotype
MIM: 610118
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR33 PubMed Links

Gene Details

Gene
GPR33
Gene Name
G protein-coupled receptor 33 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197184.2 423 Missense Mutation GAT,GGT D123G NP_001184113.2

View Full Product Details