Product Details

SNP ID
rs111625807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41319660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCAATATCTAAGTCTACTCACCA[C/T]ACCATTTTGTCACTGGAAAGGCAGA
Phenotype
MIM: 606020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OIP5 PubMed Links

Gene Details

Gene
OIP5
Gene Name
Opa interacting protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317860.1 570 Intron NP_001304789.1
NM_007280.1 570 Silent Mutation GTA,GTG V170V NP_009211.1

View Full Product Details